Order Entry
Belgium
ContactUsLinkComponent
MagBeads FastDNA kit for soil, 50 preparations, MP Biomedicals
MagBeads FastDNA kit for soil, 50 preparations, MP Biomedicals
  116561050.
 :  
undefined
MagBeads FastDNA kit for soil, 50 preparations, MP Biomedicals
  116561050.
 :  

 

  • Pk:
    50 Tests
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Soil
  • Target molecule:
    DNA
  • Description:
    MagBeads FastDNA kit for soil

 

 

Quick Isolation of genomic DNA from soil using magnetic beads technology.

  • High quality DNA: intact, high yields and free from inhibitors
  • High throughput: compatible with a wide range of automated nucleic acid purification platforms
  • Flexibility: manual and automation mode of extraction
  • Environmentally friendly: no phenol/chloroform or other toxic chemicals

The MagBeads FastDNA™ Kit for Soil allows quick and efficient isolation of high-quality genomic DNA from soil in less than 60 minutes. Samples are placed into Lysing Matrix E tubes and used with FastPrep® Instruments from MP Biomedicals to effectively lyse within 40 seconds: host cells, Bacteria, Fungi, Viruses, Protists and other cells present in soil samples.

The kit is also compatible with most of the automated nucleic acid extraction instruments on the market or it can be operated manually. It consists of specially formulated reagents to eliminate humic acid, polysaccharides, phenolic compounds and enzyme inhibitors from soil.

It allows for extraction of highly pure genomic DNA ready for PCR, restriction digestion, electrophoresis and other desired applications.