Order Entry
Belgium
ContactUsLinkComponent
SPINeasy RNA kit for bacteria, 50 preparations, MP Biomedicals
SPINeasy RNA kit for bacteria, 50 preparations, MP Biomedicals
Catalog # 116541050.
Supplier:  MP Biomedicals
CAS Number:  
SPINeasy RNA kit for bacteria, 50 preparations, MP Biomedicals
Catalog # 116541050.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    50 Tests
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Bacterial RNA
  • Sample type:
    Bacteria
  • Target molecule:
    RNA
  • Description:
    SPINeasy RNA kit for bacteria

Specifications

About this item

Quick and convenient purification of total RNA from gram-positive and gram-negative bacteria using spin column technology.

  • The kit allows multiple samples to be processed Simultaneously and without the use of toxic substances such as phenol and chloroform
  • Total RNA of high quality and integrity can be purified typically within 40 minutes is immediately available for RT-PCR and other downstream applications

SPINeasy RNA Kit for Bacteria is a silica-membrane spin-column kit that enables quick and convenient purification of total RNA from gram-positive and gram-negative bacteria. Included in the kit is our specially formulated RNASS solution that stabilizes and protects RNA in bacteria samples.

The use of Lysis Buffer R and Lysing Matrix B in combination with FastPrep® instruments from MP Biomedicals enables highly efficient lysis of bacterial samples within seconds.