Order Entry
Belgium
ContactUsLinkComponent
Sodium chloride
Sodium chloride
Catalog # SPCMSO155-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Sodium chloride
Catalog # SPCMSO155-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :58.44 g/mol
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477