Order Entry
Belgium
Orders LinkContactUsLinkComponent
 
Urea
Supplier:  Spectrum Chemical
Certificates

2 Options Available Below

Product Details & Documents

Urea, USP, JP is used in topical dermatological products to promote rehydration of the skin. Urea preparations are also used for non-surgical removal of damaged or dead nails. All Spectrum Chemical USP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol

 

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 123,00 €
    123,00 €
    1 * 500 g
       123,00 €

  • Poly pail
    12 kg
    Log in for availability
    Log in for availability
    1 * 12 kg
    1 * 12 kg - 1 640,00 €
    1 640,00 €
    1 * 12 kg
       1 640,00 €