2-Aminophenol for synthesis, Sigma-Aldrich®
Catalog # 8.00419.9010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Fibreboard box/case with liner
- Pk:10 kg
- Seq:4158
Specifications
Specification Test Results
- Solubility:17 g/l (20 °C)
- Melting Point:172 - 174 °C (sublimed)
- Molar Mass:109.12 g/mol
- Bulk Density:600 kg/m3
- Boiling Point:150 °C (15 hPa)
- Vapor Pressure:14 hPa (153 °C)
- Flash Point:>175 °C
- Density:1.328 g/cm3 (20 °C)
- pH:6.5 - 7.5 (10 g/l, H2O, 20 °C)
- Ignition Point:190 °C
- Synonyms:2-Hydroxy aniline, 2-Amino-1-hydroxy benzene
- Hill Formula:C6H7NO
- Chemical Formula:2-(NH2)C6H4OH
Specification Test Results
About this item
:
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:109.13 g/mol
UN:2512
:GHS08
:GHS07
CAS Number:95-55-6
Formula:C₆H₇NO
MDL Number:MFCD00007690