Order Entry
Belgium
ContactUsLinkComponent
L(+)-Lysine monohydrochloride ≥98.5% FCC
L(+)-Lysine monohydrochloride ≥98.5% FCC
Supplier:  Spectrum Chemical
Certificates
L(+)-Lysine monohydrochloride ≥98.5% FCC
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (C₆H₁₄N₂O₂.HCl; Dried Basis):
    98.5 - 101.5 %
  • Optical Rotation, @ 20D:
    +20.3 to +21.5
  • Lead:
    max. 5 mg/kg
  • Loss On Drying:
    max. 1.0 %
  • Residue On Ignition:
    max. 0.2 %

Specification Test Results

Frequently Bought Together