Ethyl cellulose, 10 mPa.s NF
: Ethylcellulose
:9004-57-3
:
:MFCD00131037
:Ambient
- Environmentally Preferable:
- Assay (Ethoxy Groups: -OC₂H₅; Dried Basis):44.0 - 51.0 %
- Capillary Viscosity:8.0 - 12.0 mPa.s
- Acidity or Alkalinity:To Pass Test
- Loss On Drying:max. 3.0 %
- Residue On Ignition:max. 0.5 %
Frequently Bought Together