Magnesium sulphate heptahydrate ≥99.5% FCC
Supplier: Spectrum Chemical
New Product
Certificates
About this item
Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
CAS Number:10034-99-8
:
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785
:246.48 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (After Ignition):99.5 % min.
- Loss On Ignition:40.0 - 52.0 %
- Lead (Pb):max. 4 mg/kg
- Selenium (Se):max. 0.003 %
- Identification:To Pass Test
Specification Test Results
Frequently Bought Together