Order Entry
Belgium
ContactUsLinkComponent
Methyl-4-hydroxybenzoate sodium salt ≥95.0% NF
Irritant
Methyl-4-hydroxybenzoate sodium salt ≥95.0% NF
Supplier:  Spectrum Chemical
Certificates
Irritant
Methyl-4-hydroxybenzoate sodium salt ≥95.0% NF
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (Anhydrous Basis):
    95.0 - 102.0 %
  • Completeness of Solution:
    To Pass Test
  • pH of a 1 in 1000 Solution:
    9.5 - 10.5
  • Water:
    max. 5.0 %
  • Chloride:
    max. 0.035 %

Specification Test Results

Related Information

Frequently Bought Together