ContactUsLinkComponent
HH83 UHF ACD, RFID Scanners, NFC/UHF, Handheld
HH83 UHF ACD, RFID Scanners, NFC/UHF, Handheld
Supplier:  Brady
Certificates
undefined
HH83 UHF ACD, RFID Scanners, NFC/UHF, Handheld
Supplier:  Brady
Specifications
  • Catalog No:
  • BRDY310381
  • BRDY310380
  • Memory:
  • 2 GB RAM, 16 GB Flash, MicroSD slot with support for SDXC cards
  • 2 GB RAM, 16 GB Flash, MicroSD slot with support for SDXC cards
  • W×D×H:
  • 83×133×212 mm
  • 83×133×212 mm
  • Weight:
  • 515 g
  • 515 g