Order Entry
Belgium
ContactUsLinkComponent
Respirators with FR face seal, Z-Link Series
Respirators with FR face seal, Z-Link Series
Catalog # RPBS16-010-11-CE
Supplier:  RPB SAFETY
undefined
Respirators with FR face seal, Z-Link Series
Catalog # RPBS16-010-11-CE
Supplier:  RPB SAFETY

Specifications

  • Description:
    Z-Link headtop with FR face seal and PX5 PAPR breathing tube, nylon injection molded parts, PU silencer foam, MDPE head suspension, PC safety lens, PS impact absorber, PP visor, Tychem 2000 cape/face seal, Tychem 4000 cape/face seal (sealed seams), Zytec cape/face seal/chin seal, 0,95 kg
  • Seq:
    1
  • Pk:
    1

Specifications

About this item

The RPB Z-link respirator with FR face seal, is the world's most comfortable (fully padded) and versatile industry respirator. Providing the highest respiratory protection available (TH3), the Z-Link system also provides hard hat protection, eye and face protection and hearing protection.

  • Visor hinges open for easy communication with team (outside of respiratory hazards)
  • Air flow and distribution is fully adjustable with the air deflector system to suit personal preferences
  • Fixed jaw ensures secure cape attachment
  • Vision-Link™ light mount point
  • ANSI Z89.1 certified head protection (EN 397)
  • Padded interior with adjustable ratchet system ensures a perfect customized fit that supports movement, increasing comfort and productivity
  • Three cape materials available: Tychem® 2000, Tychem® 4000, or Zytec® FR
  • Operating temperature: −10 to +60 °C
  • Noise rating: NRR 25 (side pads), NRR 41 (ear defenders and earplugs)

Optimum vision and high velocity impact protection with large optically-clear safety lens (replaceable spare part). Choice of 5 antifog and scratch resistant ANSI Z87+ (EN 166) lenses: premium clear, green tint (shade 5), yellow tint, black tint and mirror tint.

The Z-link is designed to be able to perform all tasks within a business, including general RPE, welding, grinding, painting/coating and now high heat foundry works. This allows companies to standardize systems and reduce alternate system and brands spare part stocking and usage.

To achieve this, the Z-link offers a wide range of market leading out of the box accessory items to ensure user comfort, safety and productivity. These include:
• Clip on weld visor with 1:1:1 ADF (16-670)
• Chin mounted helmet light (16-901)
• In-helmet communication system (16-922)
• Quiet-Link ear defenders (18-533)
• Comfort fit side pads (as standard)
• IR5 visors (16-810-IR5, 16-675-IR5)
• High heat foundry kits, including gold visors (16-674, 16-744, 04-955, 16-675-GT)

Certifications: EN 12941:1998+A2:2008 TH3/2 P R SL, EN 397:2012+A1:2012, EN 166:2001 Class 1BT 9 3.
Delivery: Includes PX5 PAPR breathing tube. PAPR sold separately.

Breathing tubes

Product Information