ContactUsLinkComponent
Double connector, click-on in-line super clean®
Double connector, click-on in-line super clean®
Catalog # REST22479
Supplier:  Restek
Double connector, click-on in-line super clean®
Catalog # REST22479
Supplier:  Restek
Specifications
  • Description:
    Click-on in-line super clean double connector, stainless steel
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Double connectors are used to effectively connect any click-on trap to a click-on indicator.
  • Stainless steel construction
Each double connector is 80 mm in length.