Order Entry
Belgium
ContactUsLinkComponent
MacConkey agar with CV, NaCl, and 0,15% Bile Salts, dehydrated culture media
MacConkey agar with CV, NaCl, and 0,15% Bile Salts, dehydrated culture media
Catalog # HIMEM081A-500G
Supplier:  HIMEDIA
CAS Number:  
MacConkey agar with CV, NaCl, and 0,15% Bile Salts, dehydrated culture media
Catalog # HIMEM081A-500G
Supplier:  HIMEDIA
CAS Number:  

Specifications

  • Pk:
    500 g
  • Description:
    MacConkey agar with CV, NaCl, and 0,15% Bile Salts
  • Seq:
    01

Specifications

About this item

Used to identify Enterobacteriaceae in the presence of coliforms and lactose nonfermenters from water, sewage, and food products.

  • Microbiological culture media
  • 500 g poly bottles