Order Entry
Belgium
ContactUsLinkComponent
Ethanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Ethanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC65758-25L
   Safer Human and Environmental Health
CAS Number:  
Ethanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC65758-25L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    25 L
  • Seq:
    5327
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

About this item

 :
 :46.07 g/mol
Boiling Pt:78…79 °C (1.013 hPa)
Melting Pt:–115 °C
Density:0.790…0.791 g/cm3 (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568