Hydrochloric acid, liquid, Fluka™
Box, 1
Catalog # HONC32033-1EA
Supplier: Honeywell Chemicals
Hydrochloric acid, liquid, Fluka™
Catalog # HONC32033-1EA
Supplier: Honeywell Chemicals
CAS Number: 7647-01-0
Jump to:
Product Details & Documents
Synonyms: Hydrochloric acid solution, Muriatic acid
- CAS Number:7647-01-0
- Formula:HCl
- MDL Number:MFCD00011324
- Molecular Weight:36.46 g/mol
- Storage Temperature:Ambient
- UN:3264
- ADR:8,III
Specifications
Specifications
- Pack type:Box
- Pk:1



