Manual stereotaxic injector
STOE51639
- Volume:1-100 µL
- Description:Manual stereotaxic injector
- Pk:1
Manual stereotaxic injector provides an economical means of injecting/withdrawing a calibrated volume with simple, precise manual control.
Mounted directly on a stereotaxic frame, it allows precise injection directly through the needle into the tissue. Injection directly through a needle on the syringe avoids the compliance issues that arise when injections are forced through plastic tubing. Volumes as small as 200 picoliters may be infused or withdrawn.