ContactUsLinkComponent
GMP Covers, Cylidrical Shaped with Adjustable Tyvek Drawstring Closure, Grey Tyvek®
GMP Covers, Cylidrical Shaped with Adjustable Tyvek Drawstring Closure, Grey Tyvek®
Catalog # 129-2832
undefined
GMP Covers, Cylidrical Shaped with Adjustable Tyvek Drawstring Closure, Grey Tyvek®
Catalog # 129-2832
Specifications
  • Colour:
    Grey
  • Description:
    GMP covers, Tyvek® cylindrical shaped with adjustable Tyvek drawstring closure, Ø×H 1524×304,8 mm
  • Pk:
    25
Specifications
About this item
These Grey Tyvek covers block UV light, which can damage light sensitive biotech products. Use in any pharmaceutical manufacturing setting to keep cleaned equipment protected from contamination.
  • Manufactured and packaged in a controlled cleanroom environment
  • Blocks UV Light
  • Reduces labour costs
  • Simplifies the conformance to SOP's
  • Individually poly bagged and labelled
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Grey UV Light blocking Tyvek Covers can be used in all pharmaceutical manufacturing environments to helps minimize microbial and particulate contamination risks.
Certifications: Certificates of Conformance sent with each shipment.