Aflatoxin Mix 4-solution 2 µg/mL B1+G1 and 0.5 µg/mL B2+G2 in acetonitrile, Analytical standard, reference material, Fluka™
Catalog # HONCR1961-2ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:2 mL
- Standards:Analytical standard
- Description:Aflatoxin Mix 4-solution 2 µg/mL B1+G1 and 0.5 µg/mL B2+G2 in acetonitrile
- Storage conditions:-20°C
- MDL No.:MFCD04974643
- UN No.:1648
- Concentration:2 µg/mL B1+G1 and 0.5 µg/mL B2+G2 in acetonitrile
- Description:Aflatoxin Mix 4
- Flash point:5 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery