Aflatoxin B1-solution 2 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
HONCR1956-2ML
:
- Pack type:Glass Bottle
- Pk:2 mL
- Standards:Analytical standard
- Description:Aflatoxin B1-solution 2 μg/mL in acetonitrile
- Storage conditions:-20°C
- MDL No.:MFCD00869647
- UN No.:1648
- Synonym(s):(3S,7R)-11-methoxy-6,8,19-trioxapentacyclo[10.7.0.02,9.03,7.013,17]nonadeca-1,4,9,11,13(17)-pentaene-16,18-dione
- Formula:C₁₇H₁₂O₆
- Concentration:2 μg/mL in acetonitrile
- Description:Aflatoxin B1
- Flash point:6 °C
- Physical form:Liquid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery