Orders LinkContactUsLinkComponent
Generators, Polytron® PT-DA/BEC (BIOTRONA)
Generators, Polytron® PT-DA/BEC (BIOTRONA)
Supplier:   KINEMATICA
Generators, Polytron® PT-DA/BEC (BIOTRONA)
Supplier:  KINEMATICA
Certificates
Generators, Polytron® PT-DA/BEC (BIOTRONA)
Generators, Polytron® PT-DA/BEC (BIOTRONA)
3 Options Available Below
Product Details & Documents
BIOTRONA® design high turbulence mixing. High-turbulence mixing head with low shear forces and energy inputs. Time saving with improved results compared to stirrers and dissolvers of up to 90%. Dispersing generators are made from high-alloy stainless steel 316 L, electropolished, Ra≤1,6 μm as standard.
  • Fast dissolving and suspending of solid particles into liquids
  • Ideal for higher viscosities
  • Lower shear forces
Product Family Options
Product Information
  • For
    Length
    Volume
    Availability
    Price
  • KINE11030034
    Polytron® 7 mm generator, PT-DA 07/BEC-E107 (BIOTRONA)
    Polytron® 7 mm generator, PT-DA 07/BEC-E107 (BIOTRONA)
    PT 1200 E, PT 1300 D, PT 2500 E
    107 mm
    10 - 100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 950,00 €
    1 950,00 €
    1 * 1 ST
       1 950,00 €
  • KINE11030286
    Polytron® 12 mm generator, PT-DA 12/BEC-E157 (BIOTRONA)
    Polytron® 12 mm generator, PT-DA 12/BEC-E157 (BIOTRONA)
    PT 1200 E, PT 1300 D, PT 2500 E
    157 mm
    50 - 500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 240,00 €
    2 240,00 €
    1 * 1 ST
       2 240,00 €
  • KINE11030291
    Polytron® 25 mm generator, PT-DA 25/BEC-E192 (BIOTRONA)
    Polytron® 25 mm generator, PT-DA 25/BEC-E192 (BIOTRONA)
    PT 2500 E
    192 mm
    100 - 2500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 510,00 €
    2 510,00 €
    1 * 1 ST
       2 510,00 €