Orders LinkContactUsLinkComponent
Blower transition adapters
Supplier:   LABCONCO
Blower transition adapters
Supplier:  LABCONCO
Certificates
Blower transition adapters
Blower transition adapters
2 Options Available Below
Product Details & Documents
Epoxy-coated steel blower transition adapters fit all Labconco coated steel blowers.
Adapter allows you to connect thermoplastic duct to the exhaust side of a blower to create an exhaust stack. A nominal size PVC duct fits inside the opening.
Product Family Options
Product Information
  • Weight
    Availability
    Price
  • LBCP4722401
    Blower transition adapter, 152,4 mm nominal diameter
    Blower transition adapter, 152,4 mm nominal diameter
    1,3 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • LBCP4861700
    Blower transition adapter, 254,0 mm nominal diameter
    Blower transition adapter, 254,0 mm nominal diameter
    1,4 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST