Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-Element ICP Standard, Certified Reference Material

Multi-Element ICP Standard, Certified Reference Material

ICP Standard (CRM), 100 mg/L, 2% HNO₃, 23 components, 100 mL
Catalog # HONCE2215-100ML
Supplier:   Honeywell Chemicals
Multi-Element ICP Standard, Certified Reference Material
Catalog # HONCE2215-100ML
CAS Number:  
Multi-Element ICP Standard, Certified Reference Material

ICP Standard (CRM), 100 mg/L, 2% HNO₃, 23 components, 100 mL

CAS Number:  
Price
323,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

ICP Standard Solution, 23 Components

Contents:100 mg/L each of As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, P, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn in HNO3 2%

Specifications

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Description:
    ICP Standard (CRM)
  • Contents:
    23 components

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.