ContactUsLinkComponent
I-300 compatibility kit V-7X0
I-300 compatibility kit V-7X0
Catalog # BUCH11I300S40
Supplier:  BUCHI
I-300 compatibility kit V-7X0
Catalog # BUCH11I300S40
Supplier:  BUCHI
Specifications
  • Description:
    Compatibility kit V-7X0 with 4,3" display
  • Pk:
    1
Specifications
About this item
Interface I-300 for use with BUCHI Rotavapour® R-21x/V-7x0 models.
  • Control all process parameters from single controller
  • Central control of all process parameters such as rotation speed, temperature (heating, cooling, vapour) and pressure
  • Remote monitoring on smart phones including push notifications and live view
The Interface I-300 controls all distillation parameters of a Rotavapour system. Modes: manual, timer, cont., AutoDest, drying. Mobile app is available.
Features are: Interface: I-300 Compatibility Pack: R-210/R-215, V-700/V-710.