ContactUsLinkComponent
Acetanilide Melting Point Standard, Reference Standard
Acetanilide Melting Point Standard, Reference Standard
  1004001.
 :  USP
 :  
Acetanilide Melting Point Standard, Reference Standard
  1004001.
 :  USP
 :  
  • Pack type:
    Vial
  • Pk:
    1 g
  • Standards:
    USP reference standard
  • CAS No.:
    103-84-4
  • Description:
    Acetanilide Melting Point Standard
  • Description:
    Acetanilide
  • Melting pt.:
    ~114 °C
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
 : US Pharmacopea Reference Standard