Order Entry
Belgium
Orders LinkContactUsLinkComponent
Triamcinolone Related Compounds, Reference Standard

Triamcinolone Related Compounds, Reference Standard

Supplier:   USP
Triamcinolone Related Compounds, Reference Standard
Supplier:  USP
Certificates
Triamcinolone Related Compounds, Reference Standard

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1679020.
  • 1679037.
  • Description:
  • Triamcinolone Acetonide Related Compound B
  • Triamcinolone Acetonide Related Compound C
  • Synonym(s):
  • 9-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-(11β,16α)-pregna-1,4,14-triene-3,20-dione
  • 9-Fluoro-11,21,21-trihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-(11β,16α)-pregna-1,4-diene-3,20-dione
  • CAS No.:
  • 1260149-96-9
  • 161740-69-8

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1679020.
    Triamcinolone Acetonide Related Compound B
    Triamcinolone Acetonide Related Compound B
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 240,00 €
    1 240,00 €
    1 * 15 mg
       1 240,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1679020.
    Description
    Triamcinolone Acetonide Related Compound B
    Synonym(s)
    9-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-(11β,16α)-pregna-1,4,14-triene-3,20-dione
    CAS No.
    1260149-96-9
  • 1679037.
    Triamcinolone Acetonide Related Compound C
    Triamcinolone Acetonide Related Compound C
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 270,00 €
    1 270,00 €
    1 * 15 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1679037.
    Description
    Triamcinolone Acetonide Related Compound C
    Synonym(s)
    9-Fluoro-11,21,21-trihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-(11β,16α)-pregna-1,4-diene-3,20-dione
    CAS No.
    161740-69-8