Order Entry
Belgium
ContactUsLinkComponent
 
N,N-Dimethylformamide ≥99.9% (by GC), CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # HONC34699-18L
CAS Number:  
N,N-Dimethylformamide ≥99.9% (by GC), CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # HONC34699-18L
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can, returnable
  • Pk:
    18 L
  • Seq:
    5267

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.03 %
  • Refractive index (n 20/D):
    1.4280 - 1.4310
  • Transmittance at 270 nm:
    Min. 10 %
  • Transmittance at 275 nm:
    Min. 50 %
  • Transmittance at 295 nm:
    Min. 80 %
  • Transmittance at 310 nm:
    Min. 89 %
  • Transmittance from 340 nm:
    Min. 98 %
  • RefractiveIndex:
    n20/D1.430 (lit.)
  • VaporPressure:
    2.7mmHg(20°C)
  • ExplosionLimit:
    15.2 %

Specification Test Results

About this item

 :
 :73.09 g/mol
Boiling Pt:152 - 153 °C (1,013 hPa)
Boiling Pt:152…153 °C (1,013 hPa)
Melting Pt:-61 °C
Melting Pt:–61 °C
Density:0.94 g/cm3 (20 °C)
Flash Pt:57.5 °C.1,013.25 hPa
Storage Temperature:Ambient
UN:2265
ADR:3,III
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284