Bisacodyl Related Compounds, Reference Standard
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1074030.
- 1074042.
- 1074051.
- 1074073.
- Description:
- Bisacodyl Related Compound A
- Bisacodyl Related Compound B
- Bisacodyl Related Compound C
- Bisacodyl Related Compound E
- Synonym(s):
- 4,4'-(Pyridin-2-ylmethylene)diphenol
- 2,4'-(Pyridin-2-ylmethylene)diphenol
- 4-[(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl acetate
- 2-[(4-Acetoxyphenyl)(pyridin-2-yl)methyl]phenyl acetate
- CAS No.:
- 603-41-8
- 16985-05-0
- 72901-16-7
- 111664-35-8
- Formula:
- C₁₈H₁₅NO₂
- C₁₈H₁₅NO₂
- C₂₀H₁₇NO₃
- C₂₂H₁₉NO₄
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1074030.Bisacodyl Related Compound ABisacodyl Related Compound AUSP reference standardVial20 mgLog in for availability1 * 20 mg1 180,00 €Specifications:
More
Specifications
Cat. No.1074030.DescriptionBisacodyl Related Compound ASynonym(s)4,4'-(Pyridin-2-ylmethylene)diphenolCAS No.603-41-8FormulaC₁₈H₁₅NO₂ - 1074042.Bisacodyl Related Compound BBisacodyl Related Compound BUSP reference standardVial20 mgLog in for availability1 * 20 mg1 150,00 €Specifications:
More
Specifications
Cat. No.1074042.DescriptionBisacodyl Related Compound BSynonym(s)2,4'-(Pyridin-2-ylmethylene)diphenolCAS No.16985-05-0FormulaC₁₈H₁₅NO₂ - 1074051.Bisacodyl Related Compound CBisacodyl Related Compound CUSP reference standardVial20 mgLog in for availability1 * 20 mg1 270,00 €Specifications:
More
Specifications
Cat. No.1074051.DescriptionBisacodyl Related Compound CSynonym(s)4-[(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl acetateCAS No.72901-16-7FormulaC₂₀H₁₇NO₃ - 1074073.Bisacodyl Related Compound EBisacodyl Related Compound EUSP reference standardVial20 mgLog in for availability1 * 20 mg1 200,00 €Specifications:
More
Specifications
Cat. No.1074073.DescriptionBisacodyl Related Compound ESynonym(s)2-[(4-Acetoxyphenyl)(pyridin-2-yl)methyl]phenyl acetateCAS No.111664-35-8FormulaC₂₂H₁₉NO₄



