Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pyrazinamide, Reference Standards

Pyrazinamide, Reference Standards

Supplier:   USP
Pyrazinamide, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1585006.
  • 1585017.
  • 1585028.
  • Description:
  • Pyrazinamide
  • Pyrazinamide Related Compound A
  • Pyrazinamide Related Compound B
  • Synonym(s):
  • Pyrazinecarboxylic acid
  • Pyrazinecarbonitrile
  • CAS No.:
  • 98-96-4
  • 98-97-5
  • 19847-12-2

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1585006.
    Pyrazinamide
    Pyrazinamide
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 353,00 €
    353,00 €
    1 * 200 mg
       353,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1585006.
    Description
    Pyrazinamide
    Synonym(s)
    CAS No.
    98-96-4
  • 1585017.
    Pyrazinamide Related Compound A
    Pyrazinamide Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 070,00 €
    1 070,00 €
    1 * 25 mg
       1 070,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1585017.
    Description
    Pyrazinamide Related Compound A
    Synonym(s)
    Pyrazinecarboxylic acid
    CAS No.
    98-97-5
  • 1585028.
    Pyrazinamide Related Compound B
    Pyrazinamide Related Compound B
    USP reference standard
    Ampule
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 020,00 €
    1 020,00 €
    1 * 50 mg
       1 020,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1585028.
    Description
    Pyrazinamide Related Compound B
    Synonym(s)
    Pyrazinecarbonitrile
    CAS No.
    19847-12-2
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.