Order Entry
Belgium
ContactUsLinkComponent
Ginseng Extract, Reference Standards
Ginseng Extract, Reference Standards
Supplier:  USP
Certificates
Ginseng Extract, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    90045-38-8
  • Description:
    American Ginseng
  • Formula:
    C₄₂H₆₆O₁₇

Specifications

Frequently Bought Together