Plastic Additive, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Plastic Additive 1 (Ethylene bis[3,3-bis[3-(1,1-dimethylethyl)-4-hydroxyphenyl] butanoate]) CAS 32509-66-3
Plastic Additive 2 (pentaerythrityl tetrakis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]) CAS 6683-19-8
Plastic Additive 3 (2,2',2'',6,6',6''-hexa-tert-butyl-4,4',4''-[(2,4,6-trimethyl-1,3,5-benzenetriyl)trismethylene]triphenol) CAS 1709-70-2
Plastic Additive 4 (Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate) CAS 2082-79-3
Plastic Additive 5 (Tris(2,4-di-tert-butylphenyl) phosphite) CAS 31570-04-4
Plastic Additive 6 (1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-s-triazine-2,4,6(1H,3H,5H)-trione) CAS 27676-62-6
Plastic Additive 8 (Dioctadecyl disulfide) CAS 2500-88-1
Plastic Additive 9 (Didodecyl 3,3'-thiodipropionate) CAS 123-28-4
Plastic Additive 10 (dioctadecyl 3,3'-thiodipropionate) CAS 693-36-7
Plastic Additive 11 (Copolymer of Dimethyl Succinate and (4-hydroxy-2,2,6,6-tetramethylpiperidin-1-yl)ethanol)) CAS 65447-77-0
Plastic Additive 12 (Oleamide) CAS 301-02-0
Plastic Additive 13 (Erucamide) CAS 112-84-5
Plastic Additive 14 (Di(2-ethylhexyl) phthalate) CAS 117-81-7
Plastic Additive 15 (Epoxidized Soya Oil) CAS 8013-07-8
Plastic Additive 16 (Epoxidized Linseed Oil) CAS 8016-11-3
: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Plastic Additive 1 (Ethylene bis[3,3-bis[3-(1,1-dimethylethyl)-4-hydroxyphenyl] butanoate]) CAS 32509-66-3
Plastic Additive 2 (pentaerythrityl tetrakis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]) CAS 6683-19-8
Plastic Additive 3 (2,2',2'',6,6',6''-hexa-tert-butyl-4,4',4''-[(2,4,6-trimethyl-1,3,5-benzenetriyl)trismethylene]triphenol) CAS 1709-70-2
Plastic Additive 4 (Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate) CAS 2082-79-3
Plastic Additive 5 (Tris(2,4-di-tert-butylphenyl) phosphite) CAS 31570-04-4
Plastic Additive 6 (1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-s-triazine-2,4,6(1H,3H,5H)-trione) CAS 27676-62-6
Plastic Additive 8 (Dioctadecyl disulfide) CAS 2500-88-1
Plastic Additive 9 (Didodecyl 3,3'-thiodipropionate) CAS 123-28-4
Plastic Additive 10 (dioctadecyl 3,3'-thiodipropionate) CAS 693-36-7
Plastic Additive 11 (Copolymer of Dimethyl Succinate and (4-hydroxy-2,2,6,6-tetramethylpiperidin-1-yl)ethanol)) CAS 65447-77-0
Plastic Additive 12 (Oleamide) CAS 301-02-0
Plastic Additive 13 (Erucamide) CAS 112-84-5
Plastic Additive 14 (Di(2-ethylhexyl) phthalate) CAS 117-81-7
Plastic Additive 15 (Epoxidized Soya Oil) CAS 8013-07-8
Plastic Additive 16 (Epoxidized Linseed Oil) CAS 8016-11-3
: US Pharmacopea Reference Standard
- Catalog No:
- 1544938.
- 1544949.
- 1544950.
- 1544964.
- 1544971.
- 1545056.
- 1545012.
- 1545001.
- 1545034.
- 1545045.
- 1544927.
- 1544993.
- 1545023.
- 1545067.
- 1545078.
- 1545090.
- Description:
- Plastic Additive 2
- Plastic Additive 3
- Plastic Additive 4
- Plastic Additive 5
- Plastic Additive 6
- Plastic Additive 14
- Plastic Additive 10
- Plastic Additive 9
- Plastic Additive 12
- Plastic Additive 13
- Plastic Additive 1
- Plastic Additive 8
- Plastic Additive 11
- Plastic Additive 15
- Plastic Additive 16
- Plastic Additive 18
- CAS No.:
- 64685-81-0
- UN No.:
- 3082
Product Family Options
- DescriptionStandardsPk InfoPack typePk
- Plastic Additive 2USP reference standardVial200 mg1 * 200 mg378,00 €
Specifications
Cat. No.1544938.DescriptionPlastic Additive 2CAS No.UN No. - Plastic Additive 3USP reference standardVial200 mg1 * 200 mg375,00 €
Specifications
Cat. No.1544949.DescriptionPlastic Additive 3CAS No.UN No. - Plastic Additive 4USP reference standardVial200 mg1 * 200 mg378,00 €
Specifications
Cat. No.1544950.DescriptionPlastic Additive 4CAS No.UN No. - Plastic Additive 5USP reference standardVial200 mg1 * 200 mg383,00 €
Specifications
Cat. No.1544964.DescriptionPlastic Additive 5CAS No.UN No. - Plastic Additive 6USP reference standardVial200 mg1 * 200 mg378,00 €
Specifications
Cat. No.1544971.DescriptionPlastic Additive 6CAS No.UN No. - Plastic Additive 14USP reference standardAmpule100 mg1 * 100 mg378,00 €
Specifications
Cat. No.1545056.DescriptionPlastic Additive 14CAS No.UN No. - Plastic Additive 10USP reference standardVial100 mg1 * 100 mg375,00 €
Specifications
Cat. No.1545012.DescriptionPlastic Additive 10CAS No.UN No. - Plastic Additive 9USP reference standardVial100 mg1 * 100 mg366,00 €
Specifications
Cat. No.1545001.DescriptionPlastic Additive 9CAS No.UN No. - Plastic Additive 12USP reference standardVial100 mg1 * 100 mg397,00 €
Specifications
Cat. No.1545034.DescriptionPlastic Additive 12CAS No.UN No. - Plastic Additive 13USP reference standardVial100 mg1 * 100 mg382,00 €
Specifications
Cat. No.1545045.DescriptionPlastic Additive 13CAS No.UN No. - Plastic Additive 1USP reference standardVial100 mg1 * 100 mg378,00 €
Specifications
Cat. No.1544927.DescriptionPlastic Additive 1CAS No.UN No. - Plastic Additive 8USP reference standardVial100 mg1 * 100 mg378,00 €
Specifications
Cat. No.1544993.DescriptionPlastic Additive 8CAS No.UN No. - Plastic Additive 11USP reference standardVial100 mg1 * 100 mg366,00 €
Specifications
Cat. No.1545023.DescriptionPlastic Additive 11CAS No.UN No. - Plastic Additive 15USP reference standardAmpule100 mg1 * 100 mg361,00 €
Specifications
Cat. No.1545067.DescriptionPlastic Additive 15CAS No.UN No. - Plastic Additive 16USP reference standardAmpule100 mg1 * 100 mg357,00 €
Specifications
Cat. No.1545078.DescriptionPlastic Additive 16CAS No.UN No. - Plastic Additive 18USP reference standard5 x 25 mgVial125 mg1 * 125 mg236,00 €
Specifications
Cat. No.1545090.DescriptionPlastic Additive 18CAS No.64685-81-0UN No.3082



