Pilocarpine, Reference Standards
À propos de cet article
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options disponibles ci-dessous
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Spécifications de la famille de produits
Spécifications
Spécifications
- Catalog No:
- 1538902.
- 1539009.
- Description:
- Pilocarpine Hydrochloride
- Pilocarpine Nitrate
- CAS No.:
- 54-71-7
- 148-72-1
- UN No.:
- 1544
- 1544
Spécifications
Product Family Options
Informations sur le produit
- StandardsPack typePkAvailabilityPrice
- 1538902.Pilocarpine HydrochloridePilocarpine HydrochlorideUSP reference standardVial200 mgConnexion Pour la disponibilité1 * 200 mg405,00 €Spécifications :
More
Specifications
Cat. No.1538902.DescriptionPilocarpine HydrochlorideCAS No.54-71-7UN No.1544 - 1539009.Pilocarpine NitratePilocarpine NitrateUSP reference standardVial200 mgConnexion Pour la disponibilité1 * 200 mg393,00 €Spécifications :
More
Specifications
Cat. No.1539009.DescriptionPilocarpine NitrateCAS No.148-72-1UN No.1544



