Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phenoxybenzamine, Reference Standards

Phenoxybenzamine, Reference Standards

Supplier:   USP
Phenoxybenzamine, Reference Standards
Supplier:  USP
Certificats

3 Options disponibles ci-dessous

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Caution: Cold Shipment required

Spécifications de la famille de produits

Spécifications

Spécifications

  • Catalog No:
  • 1526007.
  • 1525999.
  • 1526030.
  • Description:
  • Phenoxybenzamine Hydrochloride
  • Phenoxybenzamine Alcohol
  • Phenoxybenzamine Nitrile
  • Synonym(s):
  • 2-[Benzyl(1-phenoxypropan-2-yl)amino]ethan-1-ol
  • 3-[Benzyl(1-phenoxypropan-2-yl)amino]propanenitrile
  • CAS No.:
  • 63-92-3
  • 101-45-1
  • UN No.:
  • 3261

Spécifications

Product Family Options

Informations sur le produit

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1526007.
    Phenoxybenzamine Hydrochloride
    Phenoxybenzamine Hydrochloride
    USP reference standard
    Vial
    250 mg
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 250 mg
    1 * 250 mg - 375,00 €
    375,00 €
    1 * 250 mg
       375,00 €

    Spécifications :
    More
    Specifications
    Cat. No.
    1526007.
    Description
    Phenoxybenzamine Hydrochloride
    Synonym(s)
    CAS No.
    63-92-3
    UN No.
    3261
  • 1525999.
    Phenoxybenzamine Alcohol
    Phenoxybenzamine Alcohol
    USP reference standard
    Ampule
    50 mg
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 50 mg
    1 * 50 mg - 1 390,00 €
    1 390,00 €
    1 * 50 mg
       1 390,00 €

    Spécifications :
    More
    Specifications
    Cat. No.
    1525999.
    Description
    Phenoxybenzamine Alcohol
    Synonym(s)
    2-[Benzyl(1-phenoxypropan-2-yl)amino]ethan-1-ol
    CAS No.
    101-45-1
    UN No.
  • 1526030.
    Phenoxybenzamine Nitrile
    Phenoxybenzamine Nitrile
    USP reference standard
    Ampule
    50 mg
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 50 mg
    1 * 50 mg - 1 380,00 €
    1 380,00 €
    1 * 50 mg
       1 380,00 €

    Spécifications :
    More
    Specifications
    Cat. No.
    1526030.
    Description
    Phenoxybenzamine Nitrile
    Synonym(s)
    3-[Benzyl(1-phenoxypropan-2-yl)amino]propanenitrile
    CAS No.
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.