Order Entry
Belgium
Orders LinkContactUsLinkComponent
Anisaldehyde, Reference Standards

Anisaldehyde, Reference Standards

Supplier:   USP
Anisaldehyde, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1036948.
  • 1036937.
  • Description:
  • o-Anisaldehyde
  • p-Anisaldehyde
  • CAS No.:
  • 135-02-4
  • 123-11-5
  • UN No.:
  • 3082

Specifications

Product Family Options

Product Information

  • Standards
    Pk Info
    Pack type
    Pk
    Availability
    Price
  • 1036948.
    o-Anisaldehyde
    o-Anisaldehyde
    USP reference standard
    3 x 0.5 mL
    Ampule
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 182,00 €
    182,00 €
    1 * 3 ST
       182,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1036948.
    Description
    o-Anisaldehyde
    CAS No.
    135-02-4
    UN No.
  • 1036937.
    p-Anisaldehyde
    p-Anisaldehyde
    USP reference standard
    3 x 500 mg
    Ampule
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 366,00 €
    366,00 €
    1 * 3 ST
       366,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1036937.
    Description
    p-Anisaldehyde
    CAS No.
    123-11-5
    UN No.
    3082