Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methenamine, Reference Standards

Methenamine, Reference Standards

Fournisseur:   USP
Methenamine, Reference Standards
Fournisseur:  USP
Certificates
Methenamine, Reference Standards

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1409003.
  • 1409502.
  • 1409604.
  • Description:
  • Methenamine
  • Methenamine Hippurate
  • Methenamine Mandelate
  • CAS No.:
  • 100-97-0
  • 5714-73-8
  • 587-23-5
  • UN No.:
  • 1328

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Disponibilité
    Prix
  • 1409003.
    Methenamine
    Methenamine
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 359,00 €
    359,00 €
    1 * 500 mg
       359,00 €

    Specifications:
    Plus
    Specifications
    Cat. No.
    1409003.
    Description
    Methenamine
    CAS No.
    100-97-0
    UN No.
    1328
  • 1409502.
    Methenamine Hippurate
    Methenamine Hippurate
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 424,00 €
    424,00 €
    1 * 200 mg
       424,00 €

    Specifications:
    Plus
    Specifications
    Cat. No.
    1409502.
    Description
    Methenamine Hippurate
    CAS No.
    5714-73-8
    UN No.
  • 1409604.
    Methenamine Mandelate
    Methenamine Mandelate
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 462,00 €
    462,00 €
    1 * 200 mg
       462,00 €

    Specifications:
    Plus
    Specifications
    Cat. No.
    1409604.
    Description
    Methenamine Mandelate
    CAS No.
    587-23-5
    UN No.