Order Entry
Belgium
Orders LinkContactUsLinkComponent
Carboxymethylcellulose, Reference Standards

Carboxymethylcellulose, Reference Standards

Supplier:   USP
Carboxymethylcellulose, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1096531.
  • 1096553.
  • 1370688.
  • Description:
  • Carboxymethylcellulose Calcium
  • Carboxymethylcellulose Sodium
  • Low-substituted Carboxymethylcellulose Sodium
  • CAS No.:
  • 9050-04-8
  • 9004-32-4
  • 9004-32-4
  • Formula:
  • C₂₃H₃₀ClN₃O
  • (C₂H₄O₃)x · (Na)x

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1096531.
    Carboxymethylcellulose Calcium
    Carboxymethylcellulose Calcium
    USP reference standard
    Vial
    1,5 g
    Log in for availability
    Log in for availability
    1 * 1,5 g
    1 * 1,5 g - 236,00 €
    236,00 €
    1 * 1,5 g
       236,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1096531.
    Description
    Carboxymethylcellulose Calcium
    CAS No.
    9050-04-8
    Formula
    C₂₃H₃₀ClN₃O
  • 1096553.
    Carboxymethylcellulose Sodium
    Carboxymethylcellulose Sodium
    USP reference standard
    Vial
    1,5 g
    Log in for availability
    Log in for availability
    1 * 1,5 g
    1 * 1,5 g - 402,00 €
    402,00 €
    1 * 1,5 g
       402,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1096553.
    Description
    Carboxymethylcellulose Sodium
    CAS No.
    9004-32-4
    Formula
    (C₂H₄O₃)x · (Na)x
  • 1370688.
    Low-substituted Carboxymethylcellulose Sodium
    Low-substituted Carboxymethylcellulose Sodium
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 311,00 €
    311,00 €
    1 * 100 mg
       311,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1370688.
    Description
    Low-substituted Carboxymethylcellulose Sodium
    CAS No.
    9004-32-4
    Formula