Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dinoprostone, Reference Standards

Dinoprostone, Reference Standards

Supplier:   USP
Dinoprostone, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Caution: Cold Shipment required

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1213103.
  • 1213136.
  • Description:
  • Dinoprostone
  • Dinoprostone Related Compound C
  • Synonym(s):
  • (E)-7-{(1R,2R,3R)-3-Hydroxy-2-[(S,E)-3-hydroxyoct-1-en-1-yl]-5-oxocyclopentyl}hept-5-enoic acid
  • CAS No.:
  • 363-24-6
  • 36150-00-2

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1213103.
    Dinoprostone
    Dinoprostone
    USP reference standard
    125 mg
    Log in for availability
    Log in for availability
    1 * 125 mg
    1 * 125 mg - 3 300,00 €
    3 300,00 €
    1 * 125 mg
       3 300,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1213103.
    Description
    Dinoprostone
    Synonym(s)
    CAS No.
    363-24-6
  • 1213136.
    Dinoprostone Related Compound C
    Dinoprostone Related Compound C
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 180,00 €
    1 180,00 €
    1 * 20 mg
       1 180,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1213136.
    Description
    Dinoprostone Related Compound C
    Synonym(s)
    (E)-7-{(1R,2R,3R)-3-Hydroxy-2-[(S,E)-3-hydroxyoct-1-en-1-yl]-5-oxocyclopentyl}hept-5-enoic acid
    CAS No.
    36150-00-2