Order Entry
Belgium
Orders LinkContactUsLinkComponent
Calcium Citrate, Reference Standard

Calcium Citrate, Reference Standard

Calcium Citrate, USP reference standard, Vial, 500 mg
Catalog # 1086414.
Supplier:   USP
Calcium Citrate, Reference Standard
Catalog # 1086414.
Supplier:  USP
CAS Number:  
Calcium Citrate, Reference Standard

Calcium Citrate, USP reference standard, Vial, 500 mg

CAS Number:  
Price
494,00 €
Connexion Pour la disponibilité
1 * 500 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Spécifications

Spécifications

  • Pack type:
    Vial
  • Pk:
    500 mg
  • Standards:
    USP reference standard
  • Description:
    Calcium Citrate
  • Cat. No.:
    1086414.
  • Formula:
    C₁₂H₁₀Ca₃O₁₄ · ₄H₂O
  • Description:
    Calcium Citrate
  • Storage conditions:
    Ambient
  • Physical form:
    Solid, powder
  • CAS No.:
    5785-44-4

Spécifications