Hand held tool and covers
About this item
The hand held tool conforms to the hand and works with a choice of M-Zone hand held mop covers to ergonomically clean and scrub surfaces for a complete sanitization of small equipment and production surfaces.
- Ergonomic cleaning for small equipment and surfaces
- Lightweight, forms to hand
- Available in polypropylene, recyclable plastic, or autoclavable polyketone adapters
- Irradiated NovaPoly™ Cover is 100% smooth knit polyester, matches traditional cleanroom wipe fabrics, and provides excellent disinfectant dispersal
- PolyMesh™ Cover is 100% polyester mesh, with foam interior, for scrubbing
- Covers compatible with alcohol and strong disinfectants and are launderable for reuse
- Autoclavable
The tool adapter is available in polypropylene, recyclable plastic, or an autoclavable polyketone. The slip-on covers are available in smooth polyester or polyester mesh fabric, to meet a variety of cleaning needs.
3 Options Available Below
Product Details & Documents
The hand held tool conforms to the hand and works with a choice of M-Zone hand held mop covers to ergonomically clean and scrub surfaces for a complete sanitization of small equipment and production surfaces.
- Ergonomic cleaning for small equipment and surfaces
- Lightweight, forms to hand
- Available in polypropylene, recyclable plastic, or autoclavable polyketone adapters
- Irradiated NovaPoly™ Cover is 100% smooth knit polyester, matches traditional cleanroom wipe fabrics, and provides excellent disinfectant dispersal
- PolyMesh™ Cover is 100% polyester mesh, with foam interior, for scrubbing
- Covers compatible with alcohol and strong disinfectants and are launderable for reuse
- Autoclavable
The tool adapter is available in polypropylene, recyclable plastic, or an autoclavable polyketone. The slip-on covers are available in smooth polyester or polyester mesh fabric, to meet a variety of cleaning needs.
Documents
Micronova Cleaning Tools for the Cannabis Processing Industry
Product Literature
TDS Hand Held Mop
Datasheet
Product Family Options
Hand held mop adapter
Product Information
- L×WMaterialAvailabilityPrice
- MICRHHT-06PPHand held tool
Mop cover
Product Information
- L×WMaterialAvailabilityPrice
- MICRMZSM68-6SPolyMesh™ slip cover, 100% polyester with mesh, foam interiorPolyMesh™ slip cover, 100% polyester with mesh, foam interior152×102 mmPolyesterLog in for availability1 * 24 ST457,00 €
- MICRMZSM6-6NSNovaPoly™ slip cover, 100% smooth knit polyester, no foam, irradiatedNovaPoly™ slip cover, 100% smooth knit polyester, no foam, irradiated152×102 mmPolyesterLog in for availability1 * 24 ST420,00 €