Order Entry
Belgium
Orders LinkContactUsLinkComponent
Adaptor, QuickConnect™

Adaptor, QuickConnect™

QuickConnect™ adaptor, 13,6×2,3 cm, Stainless steel
Catalog # CONT2758
Supplier:   CONTEC
Adaptor, QuickConnect™
Catalog # CONT2758
Supplier:  CONTEC
Adaptor, QuickConnect™
Adaptor, QuickConnect™

QuickConnect™ adaptor, 13,6×2,3 cm, Stainless steel

Price
87,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel adaptor for use with QuickTask™ mops.

  • Autoclavable at 121 °C for 30 minutes

The QuickConnect™ adaptor allows users to connect any QuickConnect™ handle to the QuickTask™ polypropylene mop frame.

Specifications
  • Description:
    QuickConnect™ adaptor
  • L×W:
    13,6×2,3 cm
  • Material:
    Stainless steel
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.