Order Entry
Belgium
ContactUsLinkComponent
 
VWR® pH 2500L, pH/mV/°C Bench Meter, with Colour Graphic Touch Screen
Catalog # 662-2380
Supplier:  VWR Collection
undefined
VWR® pH 2500L, pH/mV/°C Bench Meter, with Colour Graphic Touch Screen
Catalog # 662-2380
Supplier:  VWR Collection

Specifications

  • Temperature range:
    –50,0…+200,0 °C
  • Description:
    pH/mV/°C meter, lab use
  • Model:
    pH 2500L
  • Temperature accuracy:
    ±0,1 °C
  • Temperature resolution:
    0,1 °C
  • Connections:
    BNC for pH/mV, RCA (CINCH) for temperature
  • Data logging capacity:
    500
  • Data logging - log function:
    Single collection and series of measurements
  • Display:
    127 mm colour graphic touch screen
  • Outputs:
    USB
  • Temperature compensation:
    Auto/manual
  • mV accuracy:
    ±0,1 mV
  • mV range:
    −2,000 …+2000,0 mV
  • mV resolution:
    0,1 mV / 1 mV
  • pH accuracy:
    ±0,002 pH
  • pH calibration:
    Automatic, in 1 / 3 points
  • pH range:
    −6,000...+20,000 pH
  • pH resolution:
    0,001/0,01 pH
  • Pk:
    1

Specifications

About this item

The user friendly instrument is distinguished by a large a 127 mm colour graphic touch screen. The meter can be used for accurate measurements of pH, mV and temperature. This pH meter enables simultaneous measurements displaying and storing of two functions (pH and temperature).

  • Large five colour graphic touch screen showing both pH/mV and temperature value simultaneously
  • 1- to 3-point calibration with automatic detection of buffer solutions and NIST standard solutions
  • Storing characteristics of three electrodes enables their quick replacement
  • Data-logger for up to 500 data sets - single collection and series of measurements
  • Stabilised reading signalisation
  • Connecting with a PC by a USB connector (free PC software)
  • Up to 10 last calibration reports transfer to a PC for printout
  • Automatic or manual temperature compensation
  • Multiple languages

The pH and conductivity measurement circuits are isolated, no interference during simultaneous measurements.

Delivery: pH-2500L pH meter with glass combination pH electrode; stainless steel temperature sensor; pH 4,00, pH 7,00, pH 10,00 buffer solution, 50 ml each, electrolyte – saturated KCl – 50 ml, 5 V/1000 mA power adapter; free-standing electrode holder; USB cable for connecting with a PC; carton box for the meter with accessories.

Electrodes

Product Information