ContactUsLinkComponent
Masterflex® Cleanroom 1" Sanitary Flange to Hose Barb, Avantor®
Masterflex® Cleanroom 1" Sanitary Flange to Hose Barb, Avantor®
Certificates
undefined
Masterflex® Cleanroom 1" Sanitary Flange to Hose Barb, Avantor®
Certificates
Specifications
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Flange size:
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • 1"
  • Sterilisation:
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Regulatory approvals:
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP