Orders LinkContactUsLinkComponent
Accupod solid samplers
Accupod solid samplers
Supplier:   Sampling Systems
Accupod solid samplers
Supplier:  Sampling Systems
Certificates
Accupod solid samplers
Accupod solid samplers
4 Options Available Below
Product Details & Documents
The AccuPod solid sampler is ideal for unit dose sampling of powders and granules.
  • Suitable for sampling blend uniformity
  • Next generation in sampling technology
  • Designed to minimise sampling error
The AccuPod is perfect for use in drug development and is frequently used in sampling blend uniformity. Ideal for sampling free flowing powders and granules. Sample remains in the pod until needed.
Product Family Options
Product Information
  • Material
    Volume
    Availability
    Price
  • 300-0585
    Accupod sampler, non sterile
    Accupod sampler, non sterile
    HDPE
    1 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 193,00 €
    193,00 €
    1 * 50 ST
       193,00 €
  • 300-0586
    Accupod sampler, sterile
    Accupod sampler, sterile
    HDPE
    1 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 209,00 €
    209,00 €
    1 * 50 ST
       209,00 €
  • 300-0587
    Accupod sampler, non sterile
    Accupod sampler, non sterile
    HDPE
    5 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 193,00 €
    193,00 €
    1 * 50 ST
       193,00 €
  • 300-0588
    Accupod sampler, sterile
    Accupod sampler, sterile
    HDPE
    5 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 209,00 €
    209,00 €
    1 * 50 ST
       209,00 €