EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), No-Weigh™, Pierce™
Catalog # PIERA35391
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
- Environmentally Preferable:
- Pk Info:10 x 1 mg
Specifications
Specification Test Results
- Cell Permeability:Cell-Impermeant
- Shipping Condition:Wet Ice
- Product Line:No-Weigh™, Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Carbodiimide
- Spacer:Short
- Form:Powder
- Quantity:10 x 1 mg
- Type:Crosslinker
- Solubility:Water
Specification Test Results
About this item
Synonyms: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl
:
:191.7 g/mol
UN:2811
:GHS07
Storage Temperature:Freezer
CAS Number:25952-53-8
Formula:C₈H₁₇N₃·HCl
MDL Number:MFCD00012503