Orders LinkContactUsLinkComponent
CloneMatrix
CloneMatrix
Supplier:   Molecular Devices
CloneMatrix
Supplier:  Molecular Devices
Certificates
2 Options Available Below
Product Details & Documents
CloneMatrix is a 2,5X concentrate that serves as a matrix to produce semi-solid media for hybridoma, myeloma, CHO, HEK, and mESC.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • MLDVK8510
    CloneMatrix (1×40 ml)
    CloneMatrix (1×40 ml)
    1 Bottle
    Log in for availability
    Log in for availability
    1 * 1 Fl
    1 * 1 Fl - 266,00 €
    266,00 €
    1 * 1 Fl
       266,00 €
  • MLDVK8530
    CloneMatrix CHO (1×40 ml)
    CloneMatrix CHO (1×40 ml)
    1 Bottle
    Log in for availability
    Log in for availability
    1 * 1 Fl
    1 * 1 Fl - 637,00 €
    637,00 €
    1 * 1 Fl
       637,00 €