Order Entry
Belgium
ContactUsLinkComponent
ACK Lysing Buffer (1x), TheraPEAK™
ACK Lysing Buffer (1x), TheraPEAK™
Catalog # LONZBP10-548E
Supplier:  LONZA
CAS Number:  
ACK Lysing Buffer (1x), TheraPEAK™
Catalog # LONZBP10-548E
Supplier:  LONZA
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    ACK Lysing Buffer (1x), TheraPEAK™
  • pH value:
    7,3 - 7,6

Specifications

About this item

Ammonium-Chloride-Potassium (ACK) Lysing Buffer is used to lyse red blood cells in preparations containing white blood cells.

  • Quick and easy lysis of red blood cell
  • Isolation of white blood cells

All TheraPEAK™ Products are produced according to applicable GMP standards and follow the USP/EP guidance for cell and gene therapy raw materials. It is the end user's responsibility to ensure full compliance with local regulations in terms of safety assessments and effective removal prior to patient exposure.

TheraPEAKTM Media Products are produced according to applicable GMP raw material standards and are intended to support GMP manufacturing. TheraPEAKTM Media Products are produced at FDA registered manufacturing sites with an ISO 13485 certified quality management system.