J.T.Baker®, PCR Tubes and Caps for qPCR, 8-Strips
: Avantor
PCR 8-tube strips and 8-cap strips are made of high-quality PP resin. Clear caps are compatible with qPCR application.
- Certified free from DNase, RNase and human DNA
- White 8-tube strips
- Corresponding 8-cap strips, clear to be ordered separately
- Ideal for low volume PCR and qPCR applications
- Low profile tubes help to reduce condensation
- Validated on Biorad CFX device
PCR strips and caps are manufactured under clean room conditions according to DIN EN ISO 14644-1:2016-06.
: 120 pieces of tubes and caps.
- Catalog No:
- 43001-0033
- 43001-0034
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
Frequently Bought Together