Order Entry
Belgium
ContactUsLinkComponent
Magnesium nitrate 20 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS
Magnesium nitrate 20 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS
Catalog # 85781.154
Supplier:  VWR Chemicals
CAS Number:  
undefined
Magnesium nitrate 20 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS
Catalog # 85781.154
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:13446-18-9
Formula:Mg(NO₃)₂