Order Entry
Belgium
ContactUsLinkComponent
 
Corrosive
Palladium matrix modifier 10 g/L in nitric acid 15% [99,999% Pd(NO₃)₂], ARISTAR® for graphite furnace AAS
Catalog # 91282.154
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Palladium matrix modifier 10 g/L in nitric acid 15% [99,999% Pd(NO₃)₂], ARISTAR® for graphite furnace AAS
Catalog # 91282.154
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
Storage Temperature:Ambient
UN:3264
ADR:8,II
 :GHS05