ContactUsLinkComponent
Palladium matrix modifier 5 g/L in nitric acid 1% [99,999% Pd(NO₃)₂], ARISTAR® for graphite furnace AAS
Palladium matrix modifier 5 g/L in nitric acid 1% [99,999% Pd(NO₃)₂], ARISTAR® for graphite furnace AAS
  91281.154
 :  
undefined
Palladium matrix modifier 5 g/L in nitric acid 1% [99,999% Pd(NO₃)₂], ARISTAR® for graphite furnace AAS
  91281.154
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370
 :
 :Ambient
 :1760
 :8,III
 :GHS07