Orders LinkContactUsLinkComponent
Compact burettes, BLAUBRAND®
Compact burettes, BLAUBRAND®
Supplier:   Brand
Compact burettes, BLAUBRAND®
Supplier:  Brand
Certificates
Compact burettes, BLAUBRAND®
Compact burettes, BLAUBRAND®
3 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass, amber, class AS.
  • Removable PTFE stopcock, fast to dismantle and easy to clean
  • Simple repair - all individual components replaceable
  • Calibrated to deliver (TD, Ex)
Certifications: DIN EN ISO 385, DE-M marking
Ordering information: On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
Delivery information: Delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer.
Product Family Options
Product Information
  • Capacity
    Division
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €
  • 25 ml
    0,05 ml
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €
  • 50 ml
    0,1 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.